
3,7-Dihydroxy-2-naphthoic acid

Linear Formula:
(HO)2C10H5CO2H
CAS Number:
Molecular Weight:
204.18
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing