1,3-Dioxolane
Synonym(s):
Ethylene glycol methylene ether, Formaldehyde ethylene acetal, 1,3-Dioxolane
Empirical Formula (Hill Notation):
C3H6O2
CAS Number:
Molecular Weight:
74.08
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Format
Quality Segment
Reaction Type
Special Grade
Technique
Showing 1-4 of 4