Skip to Content
Merck
1,3,5-Trimethoxybenzene Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

1,3,5-Trimethoxybenzene

1,3,5-Trimethoxybenzene Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Phloroglucinol trimethyl ether
Linear Formula:
C6H3(OCH3)3
CAS Number:
Molecular Weight:
168.19
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique