SAB3701454
Monoclonal Anti-EBI-3-Biotin antibody produced in mouse
clone V1.4C4.22, purified immunoglobulin
Synonym(s):
Anti-EBV-induced gene 3 protein, Anti-Epstein-Barr virus-induced gene 3 protein, Anti-IL-27 subunit beta, Anti-IL-27B, Anti-Mesothelian
About This Item
Recommended Products
biological source
mouse
conjugate
biotin conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
V1.4C4.22, monoclonal
form
lyophilized powder
species reactivity
mouse
technique(s)
ELISA: suitable
immunoprecipitation (IP): suitable
neutralization: suitable
western blot: suitable
isotype
IgG2b
UniProt accession no.
storage temp.
2-8°C
Gene Information
mouse ... EBI3(50498)
Specificity
Immunogen
Features and Benefits
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2
Supplementary Hazards
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service