Skip to Content
Merck
All Photos(1)

Key Documents

SMB00956

Sigma-Aldrich

trans-Resveratrol 4′-O-β-D-glucuronide

≥95% (HPLC)

Synonym(s):

Resveratrol 4′-O-glucuronide, Resveratrol-4′-O-D-Glucuronide, trans-Resveratrol 4′-O-glucuronide, trans-Resveratrol 4′-glucuronide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20O9
CAS Number:
Molecular Weight:
404.37
UNSPSC Code:
12352201
NACRES:
NA.25

Quality Level

Assay

≥95% (HPLC)

form

solid

color

white to beige

storage temp.

2-8°C

InChI

1S/C20H20O9/c21-12-7-11(8-13(22)9-12)2-1-10-3-5-14(6-4-10)28-20-17(25)15(23)16(24)18(29-20)19(26)27/h1-9,15-18,20-25H,(H,26,27)

InChI key

CDEBVTGYVFHDMA-UHFFFAOYSA-N

General description

Trans-Resveratrol 4′-O-glucuronide is a stilbenoid glycoside often present in red wine and various foods. This compound is produced through the glucuronidation process, where the enzyme UDP-glucuronosyltransferase (UGT), specifically the UGT1A9 isoform, conjugates it with glucuronic acid. It serves as a phase II metabolite of the antioxidant trans-resveratrol.

Application

Resveratrol is a plant-derived polyphenol reported to be an anticancer, antidiabetic, anti-obesity, anti-oxidant, anti-inflammatory, and anti-AD agent, among others. Some of these beneficial properties may be indirectly related to its phase II metabolism. 3-O-b-D-glucuronide (3-OG) and 4′-O-b-D-glucuronide (4OG) metabolites are more potent than resveratrol against the growth of human adenocarcinoma Caco-2. Oral resveratrol administration results in high levels of 3-OG, 4′-OG and 3-sulfate metabolites in the colorectum of colon cancer patients.

Biochem/physiol Actions

Current research indicates that trans-resveratrol 4′-O-β-D-glucuronide demonstrates its anticancer effects by activating adenosine receptors located on cancer cells, ultimately triggering apoptosis. It acts as an agonist of the adenosine A3 receptor, particularly in high concentrations, resulting in apoptosis in human colon cancer cells. Moreover, this compound effectively hinders colon carcinogenesis by suppressing cell proliferation and promoting apoptosis, making it a promising agent for combating colon cancer.

Other Notes

For additional information on our range of Biochemicals, please complete this form.
To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service