Skip to Content
Merck
All Photos(1)

Key Documents

1.16493

Sigma-Aldrich

Urease

(from jack beans) solution in 50% glycerol, 1000 U/ml for biochemistry EC 3.5.1.5

Synonym(s):

Urease enzyme

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.21

form

liquid

pH

6.0-7.0 (20 °C in H2O)

density

1.10-1.30 g/cm3 at 20 °C

storage temp.

2-8°C

General description

Urease is a nickel-containing enzyme found in plants, bacteria, fungi, algae, and soil. It is a hexamer of a single chain of 840 amino acid residues. Urease is localized to mature seeds.

Biochem/physiol Actions

Ureases are metalloenzymes that catalyze the hydrolysis of urea to release ammonia (a nitrogen source for growth) and carbon dioxide. In plants, ureases are essential for the bioavailability/metabolism of nitrogen in nature and defense mechanisms. They exhibit internal peptides-mediated insecticidal action and fungicidal activity.

Analysis Note

Activity (Urea; pH 8.0; 25 °C): ≥ 1000 U/ml
Spec. activity (Urea; pH 8.0; 25 °C): about 500 

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Urea in water and wastewater, enzymatic determination - Photometric determination using the indophenol blue method after cleavage of the urea

Urea in water and wastewater, enzymatic determination - Photometric determination using the indophenol blue method after cleavage of the urea

Urea in water and wastewater, enzymatic determination - Photometric determination using the indophenol blue method after cleavage of the urea

Urea in water and wastewater, enzymatic determination - Photometric determination using the indophenol blue method after cleavage of the urea

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service