Z129941
Thermolyne® fibrox-insulated heating tape
W × L 1 in. × 6 ft, output624 watts, Heavy-duty
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41102405
NACRES:
NB.76
Recommended Products
AC/DC input
120 V
manufacturer/tradename
Barnstead International BIH101-060
availability
not available in EU
parameter
482 °C max. temp. (900 °F)
W × L
1 in. × 6 ft
output
624 watts
Looking for similar products? Visit Product Comparison Guide
General description
Serpentine configured heating elements embedded in tough, pliable, braided tape that conforms to almost any shape vessel for even, direct-contact heating without hot spots. All tapes have high temperature lead wires emerging from opposite ends and separable line plugs. A temperature controller is required.
Standard tape has finely stranded resistance wires doubly insulated with braided fibrox yarns and knitted into flat tape. Heavy-duty version has an extra braided fibrox covering over the standard tape, suitable for direct contact on conductive surfaces.
Standard tape has finely stranded resistance wires doubly insulated with braided fibrox yarns and knitted into flat tape. Heavy-duty version has an extra braided fibrox covering over the standard tape, suitable for direct contact on conductive surfaces.
Legal Information
Thermolyne is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service