GF24963385
Constantan - resistance alloy, Cu55Ni45
insulated wire, 20m length, 0.025mm conductor diameter, 0.003mm insulation thickness, polyurethane insulation
Synonym(s):
CU045815
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
11101711
NACRES:
NA.23
Recommended Products
description
polyurethane insulation
thermocouple wire
form
wire (insulated)
manufacturer/tradename
Goodfellow 249-633-85
insulation thickness
0.003 mm
wire L × diam.
20 m × 0.025 mm , conductor diameter
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Application
may be used as one component of a thermocouple
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service