Merck
Protéines, mélange
Protéines, mélange application for HPLC
Matériaux
Colonne analytique
Réf. du produit
Description
Tarif
TSKgel® Butyl-NPR HPLC Column
phase C4 (butyl), L × I.D. 3.5 cm × 4.6 mm, 2.5 μm particle size
CONDITIONS
column
TSKgel Butyl-NPR, 3.5 cm × 4.6 mm I.D., 2.5 μm particles (814947)
mobile phase
[A] 0.1 M phosphate buffer, pH 7.0 plus ammonium sulfate, 2.3 M to 0 M in 12 min [B & C] = 0.1 M phosphate buffer, pH 7.0 plus ammonium sulfate, 1.8 M to 0 M in 60 min
flow rate
1 mL/min
detector
UV, 280 nm
Description
Informations légales
TSKgel is a registered trademark of Tosoh Corporation