Merck
Barbituriques
Barbituriques application for HPLC
Matériaux
Colonne analytique
Réf. du produit
Description
Tarif
Colonne HPLC SUPELCOSIL LC-ABZ
5 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
Discovery C18, 15 cm × 4.6 mm I.D., 5 μm particles (504955)
mobile phase
[A] acetonitrile; [B] 25 mM potassium phosphate buffer, pH 6.9 (25:75, A:B)
flow rate
1.5 mL/min
column temp.
ambient
detector
UV, 254 nm
injection
25 μL