Merck
  • Gestational Age and Child Development at Age Five in a Population-Based Cohort of Australian Aboriginal and Non-Aboriginal Children.
Gestational Age and Child Development at Age Five in a Population-Based Cohort of Australian Aboriginal and Non-Aboriginal Children.
Paediatric and perinatal epidemiology (2017-11-23)
Mark Hanly, Kathleen Falster, Georgina Chambers, John Lynch, Emily Banks, Nusrat Homaira, Marni Brownell, Sandra Eades, Louisa Jorm
RÉSUMÉ
Preterm birth and developmental vulnerability are more common in Australian Aboriginal compared with non-Aboriginal children. We quantified how gestational age relates to developmental vulnerability in both populations. Perinatal datasets were linked to the Australian Early Development Census (AEDC), which collects data on five domains, including physical, social, emotional, language/cognitive, and general knowledge/communication development. We quantified the risk of developmental vulnerability on ≥1 domains at age 5, according to gestational age and Aboriginality, for 97 989 children born in New South Wales, Australia, who started school in 2009 or 2012. Seven thousand and seventy-nine children (7%) were Aboriginal. Compared with non-Aboriginal children, Aboriginal children were more likely to be preterm (5% vs. 9%), and developmentally vulnerable on ≥1 domains (20% vs. 36%). Overall, the proportion of developmentally vulnerable children decreased with increasing gestational age, from 44% at ≤27 weeks to 20% at 40 weeks. Aboriginal children had higher risks than non-Aboriginal children across the gestational age range, peaking among early term children (risk difference [RD] 19.0, 95% confidence interval [CI] 16.3, 21.7; relative risk [RR] 1.91, 95% CI 1.77, 2.06). The relation of gestational age to developmental outcomes was the same in Aboriginal and non-Aboriginal children, and adjustment for socio-economic disadvantage attenuated the risk differences and risk ratios across the gestational age range. Although the relation of gestational age to developmental vulnerability was similar in Aboriginal and non-Aboriginal children, Aboriginal children had a higher risk of developmental vulnerability at all gestational ages, which was largely accounted for by socio-economic disadvantage.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Pyridinium dichromate, 98%