Merck
  • Human IDO-competent, long-lived immunoregulatory dendritic cells induced by intracellular pathogen, and their fate in humanized mice.
Human IDO-competent, long-lived immunoregulatory dendritic cells induced by intracellular pathogen, and their fate in humanized mice.
Scientific reports (2017-02-16)
Rajeev K Tyagi, Brodie Miles, Rajesh Parmar, Neeraj K Garg, Sarat K Dalai, Babak Baban, Christopher W Cutler
RÉSUMÉ
Targeting of myeloid-dendritic cell receptor DC-SIGN by numerous chronic infectious agents, including Porphyromonas gingivalis, is shown to drive-differentiation of monocytes into dysfunctional mDCs. These mDCs exhibit alterations of their fine-tuned homeostatic function and contribute to dysregulated immune-responses. Here, we utilize P. gingivalis mutant strains to show that pathogen-differentiated mDCs from primary human-monocytes display anti-apoptotic profile, exhibited by elevated phosphorylated-Foxo1, phosphorylated-Akt1, and decreased Bim-expression. This results in an overall inhibition of DC-apoptosis. Direct stimulation of complex component CD40 on DCs leads to activation of Akt1, suggesting CD40 involvement in anti-apoptotic effects observed. Further, these DCs drove dampened CD8
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-Mouse IgG (whole molecule) F(ab′)2 fragment–Alkaline Phosphatase antibody produced in sheep, affinity isolated antibody, buffered aqueous glycerol solution