Merck
A cation-exchange controlled core-shell MnS@Bi
Nanoscale (2017-07-12)
Yuhao Li, Yun Sun, Tianye Cao, Qianqian Su, Zili Li, Mingxian Huang, Ruizhuo Ouyang, Haizhou Chang, Shuping Zhang, Yuqing Miao
RÉSUMÉ
Overtreatment as a crucial modern medicine issue needs to be urgently addressed. Theranostic agents supply a unique platform and integrate multiple diagnosis and therapies to deal with this issue. In this study, a core-shell MnS@Bi
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Bismuth neodecanoate
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder