Merck
Compact light-emitting diode optical fiber immobilized TiO
The Science of the total environment (2017-10-04)
Heather O'Neal Tugaoen, Sergi Garcia-Segura, Kiril Hristovski, Paul Westerhoff
RÉSUMÉ
A key barrier to implementing photocatalysis is delivering light to photocatalysts that are in contact with aqueous pollutants. Slurry photocatalyst systems suffer from poor light penetration and require post-treatment to separate the catalyst. The alternative is to deposit photocatalysts on fixed films and deliver light onto the surface or the backside of the attached catalysts. In this study, TiO
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Acide 4-chlorobenzoïque, 99%