Merck
Application of galangin, an active component of Alpinia officinarum Hance (Zingiberaceae), for use in drug-eluting stents.
Scientific reports (2017-08-16)
Jung-Jin Lee, Ji-Hye Lee, Nam-Hui Yim, Joo-Hui Han, Jin Yeul Ma
RÉSUMÉ
In clinical pathology, stent interposition is used to treat vascular disease but can lead to restenosis. Drug-eluting stents (DES) are most commonly used to suppress restenosis but can also have side effects. Therefore, we investigated the anti-proliferative effect and its possible target in vitro and in vivo. We found that Alpinia officinarum Hance (AO) extract efficiently inhibited VSMC proliferation by arresting the transition from the G
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Galangin, autophagy inducing flavonoid