Merck
Electrochemical Water Oxidation by a Catalyst-Modified Metal-Organic Framework Thin Film.
ChemSusChem (2016-12-16)
Shaoyang Lin, Yuliana Pineda-Galvan, William A Maza, Charity C Epley, Jie Zhu, Matthew C Kessinger, Yulia Pushkar, Amanda J Morris
RÉSUMÉ
Water oxidation, a key component in artificial photosynthesis, requires high overpotentials and exhibits slow reaction kinetics that necessitates the use of stable and efficient heterogeneous water-oxidation catalysts (WOCs). Here, we report the synthesis of UiO-67 metal-organic framework (MOF) thin films doped with [Ru(tpy)(dcbpy)OH
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
2,2′-Bipyridine-5,5′-dicarboxylic acid, 97%