- Determining the Degree of Counterion Binding to Aqueous Alkylammonium Chloride Micelles by an Electrode Method.
Determining the Degree of Counterion Binding to Aqueous Alkylammonium Chloride Micelles by an Electrode Method.
Journal of oleo science (2016-09-16)
Hideko Sahara, Shigeharu Harada
PMID27628730
RÉSUMÉ
The degree of counterion binding (β) to micelles of aqueous n-alkylammonium chlorides (C
MATÉRIAUX