- International Union of Pharmacology. XLI. Compendium of voltage-gated ion channels: potassium channels.
International Union of Pharmacology. XLI. Compendium of voltage-gated ion channels: potassium channels.
Pharmacological reviews (2003-12-06)
George A Gutman, K George Chandy, John P Adelman, Jayashree Aiyar, Douglas A Bayliss, David E Clapham, Manuel Covarriubias, Gary V Desir, Kiyoshi Furuichi, Barry Ganetzky, Maria L Garcia, Stephan Grissmer, Lily Y Jan, Andreas Karschin, Donghee Kim, Sabina Kuperschmidt, Yoshihisa Kurachi, Michel Lazdunski, Florian Lesage, Henry A Lester, David McKinnon, Colin G Nichols, Ita O'Kelly, Jonathan Robbins, Gail A Robertson, Bernardo Rudy, Michael Sanguinetti, Susumu Seino, Walter Stuehmer, Michael M Tamkun, Carol A Vandenberg, Aguan Wei, Heike Wulff, Randy S Wymore
PMID14657415
RÉSUMÉ
This summary article presents an overview of the molecular relationships among the voltage-gated potassium channels and a standard nomenclature for them, which is derived from the IUPHAR Compendium of Voltage-Gated Ion Channels. The complete Compendium, including data tables for each member of the potassium channel family can be found at http://www.iuphar-db.org/iuphar-ic/.