Merck
Cholecystokinin antagonists: (R)-tryptophan-based hybrid antagonists of high affinity and selectivity for CCK-A receptors.
Journal of medicinal chemistry (1991-12-11)
J F Kerwin, F Wagenaar, H Kopecka, C W Lin, T Miller, D Witte, M Stashko, A M Nadzan
RÉSUMÉ
The intriguing structural similarities of glutamic acid based cholecystokinin (CCK) antagonists (A-64718 and A-65186) and the benzodiazepine CCK antagonist MK-329 (L-364,718) have been reported. Efforts to include the weak CCK antagonist benzotript into this construct utilizing a similar approach have resulted in a novel series of benzotript-based hybrid antagonists N alpha-(3'-quinolylcarbonyl)-(R)-tryptophan di-n-pentylamide (9, A-67396), N alpha-(4',8'-dihydroxy-2'-quinolylcarbonyl)-(R)-tryptophan di-n-pentylamide (23, A-70276), and N alpha-(3'-quinolylcarbonyl)-(R)-5'-hydroxytryptophan di-n-pentylamide (36, A-71134) which possess respectively binding affinities of 23, 21, and 11 nM for the pancreatic CCK-A receptor and which inhibit CCK8-induced amylase secretion. Compound 9 possesses a selectivity of greater than 500-fold for the pancreatic CCK-A receptor over the CCK-B receptor.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Pyrrole-2-carboxylic acid, 99%