Merck
Tolcapone: a review of its use in the management of Parkinson's disease.
CNS drugs (2005-02-09)
Gillian M Keating, Katherine A Lyseng-Williamson
RÉSUMÉ
Tolcapone (Tasmar) is a selective, reversible inhibitor of peripheral and central catechol-O-methyltransferase (COMT). Results of well designed studies indicate that oral tolcapone is an effective adjunct to levodopa plus a peripheral dopa-decarboxylase inhibitor (DDCI) in patients with fluctuating Parkinson's disease. Tolcapone significantly improves levodopa-induced motor fluctuations and significantly reduces levodopa requirements. The drug is generally well tolerated, with the most commonly occurring adverse events being dopaminergic related. Thus, tolcapone is a useful option in patients with fluctuating Parkinson's disease who are receiving levodopa/DDCI and are not responding to, or are not candidates for, other adjunctive treatments.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Tolcapone, ≥98% (HPLC)