Merck
Photoaffinity labelling of cyanomethaemoglobin with derivatives of tryptophan and 5-bromotryptophan.
The Biochemical journal (1995-05-15)
M Li, Z Lin, M E Johnson
RÉSUMÉ
Tryptophan and 5-bromotryptophan (5-BrTrp) are relatively potent inhibitors of sickle-haemoglobin polymerization. The binding sites of these compounds to normal and sickle haemoglobin (HBA and HBS) have been suggested, but not firmly established, through the use of spin-labelled derivatives and/or computer modeling. In the present study we approached the problem by utilizing the technique of photoaffinity labelling. The cyanomet forms of HBA and HBS were subjected to photoaffinity labelling with N alpha-(4-azidotetrafluorobenzoyl)tryptophan and N alpha-(1-ethyl-2-diazomalonyl)-5-bromotryptophan respectively. Both irradiated samples of HBA and HBS were denatured, digested with trypsin, and then separated by reversed-phase HPLC. A labelled tryptic peptide was isolated from the photolabelling of HBS with N alpha-(1-ethyl-2-diazomalonyl)-5-bromotryptophan. The peptide was identified to be Val1(alpha)-Lys7(alpha), with the label attached to Val1(alpha), by virtue of amino acid analysis and sequencing, in conjunction with fast-atom-bombardment MS. The binding mode of N alpha-(1-ethyl-2-diazomalonyl)-5-bromotryptophan is proposed and its relevance to the potency of the 5-BrTrp-based anti-sickling agents is discussed.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
5-Bromo-DL-tryptophan, 99%