Merck
  • [The comparison of neuroprotective action of antibodies to glutamate and drug preparation semax in the focal ischemic damage of the brain prefrontal cortex of rats].
[The comparison of neuroprotective action of antibodies to glutamate and drug preparation semax in the focal ischemic damage of the brain prefrontal cortex of rats].
Patologicheskaia fiziologiia i eksperimental'naia terapiia (2012-10-18)
G A Romanova, F M Shakova, T V Davydova
RÉSUMÉ
It was stated, that with bilateral photochemically induced thrombosis of the prefrontal cortex peptide semax and the AB-Glu by intranasal injection provoke pronounced neuroprotective and antiamnestic action. Intranasal injection semax (250 mkg/kg/daily during six postoperative days) and AB-Glu (250 mkg/kg in 1 hour after phototrombosis) demonstrate diminishing of cortex damage volume and relieve preservation and reproduction rat passive avoidance reflex, acquired before bilateral photochemically induced thrombosis of prefrontal cortex.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Semax, ≥98% (HPLC)
Le tarif et la disponibilité ne sont pas disponibles actuellement.