Merck
Microchannel liquid-flow focusing and cryo-polymerization preparation of supermacroporous cryogel beads for bioseparation.
Journal of chromatography. A (2012-06-15)
Junxian Yun, Changming Tu, Dong-Qiang Lin, Linhong Xu, Yantao Guo, Shaochuan Shen, Songhong Zhang, Kejian Yao, Yi-Xin Guan, Shan-Jing Yao
RÉSUMÉ
Polymeric cryogels are sponge-like materials with supermacroporous structure, allowing them to be of interest as new chromatographic supports, cell scaffolds and drug carriers in biological and biomedical areas. The matrices of cryogels are always prepared in the form of monoliths by cryo-polymerization under frozen conditions. However, there are limited investigations on the production of cryogels in the form of adsorbent beads suitable for bioseparation. In this work, we provide a new approach by combining the microchannel liquid-flow focusing with cryo-polymerization for the preparation of polyacrylamide-based supermacroporous cryogel beads with a narrow particle size distribution. The present method was achieved by introducing the aqueous phase solution containing monomer, cross-linker and redox initiators, and the water-immiscible organic oil phase containing surfactant simultaneously into a microchannel with a cross-shaped junction, where the aqueous drops with uniform sizes were generated by the liquid shearing and the segmentation due to the steady flow focusing of the immiscible phase streams. These liquid drops were in situ suspended into the freezing bulk oil phase for cryo-polymerization and the cryogel matrix beads were obtained by thawing after the achievement of polymerization. By grafting the polymer chains containing sulfo binding groups onto these matrix beads, the cation-exchange cryogel beads for protein separation were produced. The results showed that at the aqueous phase velocities from 0.5 to 2.0 cm/s and the total velocities of the water-immiscible phase from 2.0 to 6.0 cm/s, the obtained cryogel beads by the present method have narrow size distributions with most of the bead diameters in the range from 800 to 1500 μm with supermacropores in sizes of about 3-50 μm. These beads also have high porosities with the averaged maximum porosity of 96.9% and the mean effective porosity of 86.2%, which are close to those of the polyacrylamide-based cryogel monoliths. The packed bed using the cryogel beads with mean diameter of 1248 μm, as an example, has reasonable and acceptable liquid dispersion, but high water permeability (4.29 × 10⁻¹⁰ m²) and high bed voidage (90.2%) owing to the supermacropores within the beads, enhanced the rapid binding and separation of protein from the feedstock even at high flow velocities. The purity of the obtained lysozyme from chicken egg white by one-step chromatography using the packed bed was in the range of about 78-92% at the flow velocities of 0.5-15 cm/min, indicating that the present cryogel beads could be an effective chromatographic adsorbent for primary bioseparation.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
2-Acrylamido-2-methyl-1-propanesulfonic acid, 99%
Sigma-Aldrich
2-Acrylamido-2-methyl-1-propanesulfonic acid sodium salt solution, 50 wt. % in H2O