Merck
Development of programmable small DNA-binding molecules with epigenetic activity for induction of core pluripotency genes.
Bioorganic & medicinal chemistry (2012-03-13)
Ganesh N Pandian, Akimichi Ohtsuki, Toshikazu Bando, Shinsuke Sato, Kaori Hashiya, Hiroshi Sugiyama
RÉSUMÉ
Epigenetic modifications that govern the gene expression are often overlooked with the design of artificial genetic switches. N-Methylpyrrole-N-methylimidazole (PI) hairpin polyamides are programmable small DNA binding molecules that have been studied in the context of gene regulation. Recently, we synthesized a library of compounds by conjugating PI polyamides with SAHA, a chromatin-modifier. Among these novel compounds, PI polyamide-SAHA conjugate 1 was shown to epigenetically activate pluripotency genes in mouse embryonic fibroblasts. Here, we report the synthesis of the derivatives of conjugate 1 and demonstrate that these epigenetically active molecules could be developed to improve the induction of pluripotency factors.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
1-Methylimidazole, ReagentPlus®, 99%
Sigma-Aldrich
1-Methylimidazole, ≥99%, purified by redistillation