Merck
Studies on cerebral protection of digoxin against ischemia/reperfusion injury in mice.
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan (2009-04-02)
Shaminder Kaur, Ashish K Rehni, Nirmal Singh, Amteshwar S Jaggi
RÉSUMÉ
The present study was designed to investigate the possible neuroprotective effect of digoxin induced pharmacological preconditioning (PP) and its probable mechanism. Bilateral carotid artery occlusion (BCAO) of 17 min followed by reperfusion for 24 h was employed to produce ischemia and reperfusion (I/R) induced cerebral injury in male swiss albino mice. Cerebral infarct size was measured using triphenyltetrazolium chloride staining. Memory was assessed using elevated plus maze test. Degree of motor incoordination was evaluated using inclined beam walking test, rota rod test and lateral push test. Digoxin (0.08 mg/kg, i.p.) was administered 24 h before surgery in a separate group of animals to induce PP. BCAO followed by reperfusion, produced significant rise in cerebral infarct size along with impairment of memory and motor coordination. Digoxin treatment produced a significant decrease in cerebral infarct size and reversal of I/R induced impairment of memory and motor incoordination. Digoxin induced neuroprotective effect was abolished significantly by verapamil (15 mg/kg, i.p.), a L-type calcium channel blocker, ruthenium red (3 mg/kg, s.c.), an intracellular ryanodine receptor blocker and 3,4-dichlorobenzamil (Na(+)/Ca(2+) exchanger inhibitor). These findings indicate that digoxin preconditioning exerts a marked neuroprotective effect on the ischemic brain, which is possibly linked to digitalis induced increase in intracellular calcium levels eventually leading to the activation of calcium sensitive signal transduction cascades.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
3′,4′-Dichlorobenzamil hydrochloride, >98% (HPLC)