- Fluorescent chemosensor for reactive organohalides in micellar solution with an example of autocatalysis.
Fluorescent chemosensor for reactive organohalides in micellar solution with an example of autocatalysis.
Chemical communications (Cambridge, England) (2009-04-01)
Jung-Jae Lee, Bradley D Smith
PMID19333457
RÉSUMÉ
N-Alkylation of a fluorescent macrocyclic amine in aqueous micellar solution produces enhanced emission; the reaction with chloromethyl methyl ether exhibits autocatalysis.