Merck
  • PDE type-4 inhibition increases L-type Ca(2+) currents, action potential firing, and quantal size of exocytosis in mouse chromaffin cells.
PDE type-4 inhibition increases L-type Ca(2+) currents, action potential firing, and quantal size of exocytosis in mouse chromaffin cells.
Pflugers Archiv : European journal of physiology (2008-09-10)
A Marcantoni, V Carabelli, D H Vandael, V Comunanza, E Carbone
RÉSUMÉ
We studied the effects of the cAMP-hydrolyzing enzyme phosphodiesterase type-4 (PDE4) on the L-type Ca(2+) channels (LTCCs) and Ca(2+)-dependent secretion in mouse chromaffin cells (MCCs). The selective PDE4 inhibitor rolipram (3 microM) had a specific potentiating action on Ca(2+) currents of MCCs (40% increase within 3 min). A similar effect was produced by the selective beta(1)-AR agonist denopamine (1 microM) and by the unselective PDEs inhibitor IBMX (100 microM). Rolipram and denopamine actions were selective for LTCCs, and the Ca(2+) current increase remained unchanged if the two compounds were applied simultaneously. This suggests that at rest, LTCCs in MCCs are down-regulated by the low levels of cAMP determined by PDE4 activity and that LTCCs can be up-regulated by either inhibiting PDE4 or activating beta(1)-AR. No other PDEs are likely involved in this specific action. PDE4 inhibition had also a marked effect on the spontaneous firing of resting MCCs and catecholamine secretion. Rolipram up-regulated the LTCCs contributing to the "pace-maker" current underlying action potential (AP) discharges and accelerated the firing rate, with no significant effects on AP waveform. Acceleration of AP firing was also induced by the LTCC-agonist Bay K (1 microM), while nifedipine (3 microM) reduced the firing frequency, suggesting that LTCCs and intracellular cAMP play a key role in setting the pace-maker current regulating MCCs excitability. Rolipram increased also the size of the ready-releasable pool and the quantal content of secretory vesicles without affecting their probability of release. Thus, rolipram acts on MCCs by up-regulating both exocytosis and AP firings. These two processes are effectively down-regulated by PDE4 at rest and can dramatically increase the quantity of released catecholamines when PDE4 is inhibited and/or cAMP is raised.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
R(−)-Denopamine, ≥98% (HPLC), powder