Merck
  • The diacylglycerol lipase inhibitor RHC-80267 potentiates the relaxation to acetylcholine in rat mesenteric artery by anti-cholinesterase action.
The diacylglycerol lipase inhibitor RHC-80267 potentiates the relaxation to acetylcholine in rat mesenteric artery by anti-cholinesterase action.
European journal of pharmacology (2005-06-17)
Philippe Ghisdal, Greet Vandenberg, Marie-Christine Hamaide, Maurice Wibo, Nicole Morel
RÉSUMÉ
The diacylglycerol lipase inhibitor 1,6-bis(cyclohexyloximinocarbonylamino) hexane (RHC-80267) was tested for its effect on acetylcholine-evoked relaxation in rat mesenteric artery. In artery contracted with either noradrenaline or KCl, RHC-80267 (0.1-10 muM) potentiated the relaxation evoked by acetylcholine. The effect of RHC-80267 was not affected by nitric oxide synthase inhibition or by inhibitors of protein kinase C or of phospholipase A(2). The diacylglycerol analogue 1-oleoyl-2-acetyl-sn-glycerol did not change the relaxation to acetylcholine. RHC-80267 did not affect the relaxation evoked by carbachol, by the nitric oxide donor SNAP (S-nitroso-N-acetylpenicillamine) or by the K(+) channel opener cromakalim. Neostigmine, a cholinesterase inhibitor, produced the same effect as RHC-80267 on acetylcholine-evoked relaxation. When tested on cholinesterase in brain homogenate, RHC-80267 concentration-dependently inhibited cholinesterase activity with an IC(50) of 4 muM. These results indicate that the potentiation of acetylcholine-evoked responses by RHC-80267 in rat mesenteric artery is caused by the inhibition of the cholinesterase activity in the vascular wall.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
RHC 80267, ≥98% (HPLC), solid