Merck
ERAD inhibitors integrate ER stress with an epigenetic mechanism to activate BH3-only protein NOXA in cancer cells.
Proceedings of the National Academy of Sciences of the United States of America (2009-01-24)
Qiuyan Wang, Helena Mora-Jensen, Marc A Weniger, Patricia Perez-Galan, Chris Wolford, Tsonwin Hai, David Ron, Weiping Chen, William Trenkle, Adrian Wiestner, Yihong Ye
RÉSUMÉ
The ubiquitin-proteasome system has recently emerged as a major target for drug development in cancer therapy. The proteasome inhibitor bortezomib has clinical activity in multiple myeloma and mantle cell lymphoma. Here we report that Eeyarestatin I (EerI), a chemical inhibitor that blocks endoplasmic reticulum (ER)-associated protein degradation, has antitumor and biologic activities similar to bortezomib and can synergize with bortezomib. Like bortezomib, EerI-induced cytotoxicity requires the up-regulation of the Bcl-2 homology3 (BH3)-only pro-apoptotic protein NOXA. We further demonstrate that both EerI and bortezomib activate NOXA via an unanticipated mechanism that requires cooperation between two processes. First, these agents elicit an integrated stress response program at the ER to activate the CREB/ATF transcription factors ATF3 and ATF4. We show that ATF3 and ATF4 form a complex capable of binding to the NOXA promoter, which is required for NOXA activation. Second, EerI and bortezomib also block ubiquitination of histone H2A to relieve its inhibition on NOXA transcription. Our results identify a class of anticancer agents that integrate ER stress response with an epigenetic mechanism to induce cell death.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Eeyarestatin I, ≥98% (HPLC)