Merck
Adipocyte-specific deletion of zinc finger protein 407 results in lipodystrophy and insulin resistance in mice.
Molecular and cellular endocrinology (2020-12-08)
Alyssa Charrier, Xuan Xu, Bo-Jhih Guan, Justine Ngo, Anthony Wynshaw-Boris, Maria Hatzoglou, David A Buchner
RÉSUMÉ
PPARγ deficiency in humans and model organisms impairs the transcriptional control of adipogenesis and mature adipocyte function resulting in lipodystrophy and insulin resistance. Zinc finger protein 407 (ZFP407) positively regulates PPARγ target gene expression and insulin-stimulated glucose uptake in cultured adipocytes. The in vivo physiological role of ZFP407 in mature adipocytes, however, remains to be elucidated. Here we generated adipocyte-specific ZFP407 knockout (AZKO) mice and discovered a partial lipodystrophic phenotype with reduced fat mass, hypertrophic adipocytes in inguinal and brown adipose tissue, and reduced adipogenic gene expression. The lipodystrophy was further exacerbated in AZKO mice fed a high-fat diet. Glucose and insulin tolerance tests revealed decreased insulin sensitivity in AZKO mice compared to control littermates. Cell-based assays demonstrated that ZFP407 is also required for adipogenesis, which may also contribute to the lipodystrophic phenotype. These results demonstrate an essential in vivo role of ZFP407 in brown and white adipose tissue formation and organismal insulin sensitivity.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Zfp407