Merck
  • Therapy Resistance in Neoadjuvantly Treated Gastric Cancer and Cancer of the Gastroesophageal Junction is Associated with an Increased Expression of Immune Checkpoint Inhibitors-Comparison Against a Therapy Naïve Cohort.
Therapy Resistance in Neoadjuvantly Treated Gastric Cancer and Cancer of the Gastroesophageal Junction is Associated with an Increased Expression of Immune Checkpoint Inhibitors-Comparison Against a Therapy Naïve Cohort.
Translational oncology (2019-12-23)
Hauke Schoop, Anna Bregenzer, Christine Halske, Hans-Michael Behrens, Sandra Krüger, Jan-Hendrik Egberts, Christoph Röcken
RÉSUMÉ
With recent studies uncovering the complex landscape of immune checkpoint regulators in gastric cancer (GC), we aimed to characterize the expression of the checkpoint proteins V-domain Ig suppressor of T-cell activation (VISTA), programmed cell death 1 ligand 1 (PD-L1), and programmed cell death protein-1 (PD-1) in a cohort of GCs following platinum-based neoadjuvant chemotherapy. A total of 141 GC samples, 93 lymph node metastases, and 15 distant metastases were assessed using immunohistochemistry. Staining results were correlated with clinicopathological patient characteristics, genetic alterations, and survival. The expression of VISTA was detected in tumor cells of 38 (30.9%) GCs and immune cells of 139 (98.6%) GCs. The expression of PD-L1 was detected in tumor cells of 27 (22.7%) GCs and immune cells of 134 (96.4%) GCs. The expression of PD-1 was only observed in lymphocyte aggregates/intratumoral lymphoid follicles of 123 (87.2%) GCs. VISTA and PD-L1 correlated in their expression and were associated with poor tumor regression. Compared with an ancient cohort of therapy naïve GCs, we observed a major increase in overall immune cell density accompanied by an over proportional increase in PD-1 and VISTA-positive immune cells. The frequency of VISTA expression in tumor cells was also found to be substantially increased. To the contrary, expression of PD-L1 was decreased in immune cells and tumor cells of neoadjuvantly treated GCs. As a result, a subset of GCs using a single (only VISTA or PD-L1) or combined (VISTA and PD-L1) immune evasion mechanisms might benefit from an anti-PD-L1/anti-VISTA-targeted therapy.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anticorps anti-MSH2, clone FE11, clone FE11, from mouse