Merck
Autophagy gene Atg16L1 prevents lethal T cell alloreactivity mediated by dendritic cells.
Immunity (2014-10-14)
Vanessa M Hubbard-Lucey, Yusuke Shono, Katie Maurer, Mallory L West, Natalie V Singer, Carly G K Ziegler, Cecilia Lezcano, Ana Carolina Fragoso Motta, Karin Schmid, Samuel M Levi, George F Murphy, Chen Liu, Jeffrey D Winkler, Ravi K Amaravadi, Gerhard Rogler, Anne M Dickinson, Ernst Holler, Marcel R M van den Brink, Ken Cadwell
RÉSUMÉ
Atg16L1 mediates the cellular degradative process of autophagy and is considered a critical regulator of inflammation based on its genetic association with inflammatory bowel disease. Here we find that Atg16L1 deficiency leads to an exacerbated graft-versus-host disease (GVHD) in a mouse model of allogeneic hematopoietic stem cell transplantation (allo-HSCT). Atg16L1-deficient allo-HSCT recipients with GVHD displayed increased T cell proliferation due to increased dendritic cell (DC) numbers and costimulatory molecule expression. Reduced autophagy within DCs was associated with lysosomal abnormalities and decreased amounts of A20, a negative regulator of DC activation. These results broaden the function of Atg16L1 and the autophagy pathway to include a role in limiting a DC-mediated response during inflammatory disease, such as GVHD.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-Atg16L antibody produced in rabbit, fractionated antiserum, buffered aqueous solution