Merck
Biallelic mutations in p16(INK4a) confer resistance to Ras- and Ets-induced senescence in human diploid fibroblasts.
Molecular and cellular biology (2002-11-06)
Thomas J Huot, Janice Rowe, Mark Harland, Sarah Drayton, Sharon Brookes, Chandra Gooptu, Patricia Purkis, Mike Fried, Veronique Bataille, Eiji Hara, Julia Newton-Bishop, Gordon Peters
RÉSUMÉ
The INK4a/ARF tumor suppressor locus is implicated in the senescence-like growth arrest provoked by oncogenic Ras in primary cells. INK4a and ARF are distinct proteins encoded by transcripts in which a shared exon is decoded in alternative reading frames. Here we analyze dermal fibroblasts (designated Q34) from an individual carrying independent missense mutations in each copy of the common exon. Both mutations alter the amino acid sequence of INK4a and functionally impair the protein, although they do so to different degrees. Only one of the mutations affects the sequence of ARF, causing an apparently innocuous change near its carboxy terminus. Unlike normal human fibroblasts, Q34 cells are not permanently arrested by Ras or its downstream effectors Ets1 and Ets2. Moreover, ectopic Ras enables the cells to grow as anchorage-independent colonies, and in relatively young Q34 cells anchorage independence can be achieved without addition of telomerase or perturbation of the p53 pathway. Whereas ARF plays the principal role in Ras-induced arrest of mouse fibroblasts, our data imply that INK4a assumes this role in human fibroblasts.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Anti-MDM2 (Ab-1) Mouse mAb (IF2), liquid, clone IF2, Calbiochem®
Sigma-Aldrich
Anti-MDM2 (Ab-2) Mouse mAb (2A10), liquid, clone 2A10, Calbiochem®