Merck
ER stress causes rapid loss of intestinal epithelial stemness through activation of the unfolded protein response.
Cell reports (2013-04-03)
Jarom Heijmans, Jooske F van Lidth de Jeude, Bon-Kyoung Koo, Sanne L Rosekrans, Mattheus C B Wielenga, Marc van de Wetering, Marc Ferrante, Amy S Lee, Jos J M Onderwater, James C Paton, Adrienne W Paton, A Mieke Mommaas, Liudmila L Kodach, James C Hardwick, Daniël W Hommes, Hans Clevers, Vanesa Muncan, Gijs R van den Brink
RÉSUMÉ
Stem cells generate rapidly dividing transit-amplifying cells that have lost the capacity for self-renewal but cycle for a number of times until they exit the cell cycle and undergo terminal differentiation. We know very little of the type of signals that trigger the earliest steps of stem cell differentiation and mediate a stem cell to transit-amplifying cell transition. We show that in normal intestinal epithelium, endoplasmic reticulum (ER) stress and activity of the unfolded protein response (UPR) are induced at the transition from stem cell to transit-amplifying cell. Induction of ER stress causes loss of stemness in a Perk-eIF2α-dependent manner. Inhibition of Perk-eIF2α signaling results in stem cell accumulation in organoid culture of primary intestinal epithelium. Our findings show that the UPR plays an important role in the regulation of intestinal epithelial stem cell differentiation.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anticorps anti-"achaete scute homolog 2", clone 7E2, clone 7E2, from mouse