- BABIPhos Family of Biaryl Dihydrobenzooxaphosphole Ligands for Asymmetric Hydrogenation.
BABIPhos Family of Biaryl Dihydrobenzooxaphosphole Ligands for Asymmetric Hydrogenation.
Organic letters (2018-03-16)
Guisheng Li, Olga V Zatolochnaya, Xiao-Jun Wang, Sonia Rodríguez, Bo Qu, Jean-Nicolas Desrosiers, Hari P R Mangunuru, Soumik Biswas, Daniel Rivalti, Shuklendu D Karyakarte, Joshua D Sieber, Nelu Grinberg, Ling Wu, Heewon Lee, Nizar Haddad, Daniel R Fandrick, Nathan K Yee, Jinhua J Song, Chris H Senanayake
PMID29542928
RÉSUMÉ
Novel bidentate phosphine ligands BABIPhos featuring a biaryl bis-dihydrobenzooxaphosphole core are presented. Their synthesis was achieved via Pd-catalyzed reductive homocoupling of dihydrobenzooxaphosphole aryl triflates. An efficient route toward various analogues was also established, giving access to phosphines with different electronic and steric properties. The newly obtained ligands demonstrated high efficiency and selectivity in Rh-catalyzed asymmetric hydrogenation of di- and trisubstituted enamides. This new class of ligands is complementary to previously described bidentate benzooxaphosphole ligands BIBOP.