MilliporeSigma
Omega-3 Fatty Acids Responsive Proteins and Reduction in Breast Density in Obese Postmenopausal Women.
Journal of proteome research (2019-08-02)
Yuan-Wan Sun, Haifang Xu, Gabrielle Benitez, Kun-Ming Chen, Anne Stanley, Bruce Stanley, Junjia Zhu, Henry Thompson, Andrea Manni, Karam El-Bayoumy
RÉSUMÉ
We reported that breast density (BD) was inversely correlated with the plasma level of DHA in postmenopausal obese, but not in nonobese, women given Lovaza (n-3FA). To identify protein biomarkers for the possible differential effect of n-3FA on BD between obese and nonobese women, an iTRAQ method was performed to analyze plasma from obese and lean women at each time point (baseline, 12 and 24-months, n = 10 per group); 173 proteins with >95% confidence (Unuses Score >1.3 and local false discovery rate estimation <5%) were identified. Comparative analysis between various groups identified several differentially expressed proteins (hemopexin precursor, vitamin D binding protein isoform 1 precursor [VDBP], fibronectin isoform 10 precursor [FN], and α-2 macroglobulin precursor [A2M]). Western blot analysis was performed to verify the differential expression of proteins in the iTRAQ study, and those found to be altered in a tumor protective fashion by an n-3FA rich diet in our previous preclinical study; gelsolin, VDBP, and FN were altered by n-3FA in a manner consistent with reduction in inflammation in obese women. To test the impact of our findings on breast cancer risk reduction by n-3FA, a posthoc analysis revealed that n-3FA administration reduced BD selectively in obese postmenopausal women.