Merck
  • Impact of the metabotropic glutamate receptor7 (mGlu7) allosteric agonist, AMN082, on fear learning and memory and anxiety-like behavior.
Impact of the metabotropic glutamate receptor7 (mGlu7) allosteric agonist, AMN082, on fear learning and memory and anxiety-like behavior.
European journal of pharmacology (2019-07-02)
Jolanta H Kotlinska, Malgorzata Lopatynska-Mazurek, Kinga Gawel, Patrycja Gabka, Malgorzata Jenda-Wojtanowska, Marta Kruk-Slomka, Marta Marszalek-Grabska, Zofia Danilczuk, Ewa Kedzierska, Sylwia Talarek, Joanna Listos, Ewa Gibula-Tarlowska
RÉSUMÉ
The present study was conducted to evaluate the influence of AMN082, the metabotropic glutamate receptor subtype 7 (mGlu7) allosteric agonist on different stages of memory processes connected with fear conditioning in the passive avoidance (PA) learning task in mice and negative emotional state (anxiety-like) induced by ethanol- and morphine-withdrawal in the elevated plus maze (EPM) test in rats. To perform the PA test, AMN082 (1.25, 2.5 and 5 mg/kg, i. p.) was injected to interfere with (or inhibit) acquisition, consolidation, and retrieval processes. The retention latency in each group was recorded using a step-through passive avoidance task 24 h after training. In turn, in ethanol- and morphine-withdrawal rats, the influence of AMN082 on anxiety-like behavior was estimated in the EPM test 24 h- (ethanol) and 72- h (morphine) after the last dose of repeated drug administrations. In all experimental groups, AMN082 at the dose of 5 mg/kg significantly decreased the step-through latency of long-term memory in the PA task. These AMN082 effects were reversed by MMPIP (10 mg/kg), the antagonist of mGlu7 receptor. AMN082 (2.5 and 5 mg/kg) also decreased ethanol- and morphine withdrawal-induced anxiety-like behavior in the EPM test, and this AMN082 (5 mg/kg) effect was counteracted by MMPIP pretreatment. Taken together, the results show that mGlu7 is involved in fear learning to the context and anxiety-like state connected with unpleasant experiences after ethanol- and morphine withdrawal in rodents. However, it appears that functional dissociation exists between these two AMN082 effects.