Merck
  • Separation of synthetic oligonucleotide dithioates from monothiophosphate impurities by anion-exchange chromatography on a mono-q column.
Separation of synthetic oligonucleotide dithioates from monothiophosphate impurities by anion-exchange chromatography on a mono-q column.
Analytical biochemistry (2002-06-19)
Xianbin Yang, Richard P Hodge, Bruce A Luxon, Robert Shope, David G Gorenstein
RÉSUMÉ
A method using a strong anion-exchange liquid-chromatography column, Mono-Q, has been developed for high-resolution analysis and purification of oligonucleotide dithioates, which were synthesized by an automated, solid-phase, phosphorothioamidite chemistry. High-resolution separation of oligonucleotide phosphorodithioates from monothiophosphate impurities was obtained. High-resolution separation was also demonstrated at pH 8. The separation of oligonucleotide dithioates was found to be linearly dependent on the number of sulfurs for the same sequence length. Thiocyanate, SCN-, as eluting anion, can be used to purify oligonucleotides containing a high percentage of phosphorodithioate linkages in lower salt concentrations and provides better separation than chloride as eluting anion.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Mono Q® HR 5/5, 10 μm particle size, L × I.D. 5 cm × 5 mm