Merck
  • Assessment of the Thermodynamic Properties of DL-p-Mentha-1,8-diene, 4-Isopropyl-1-Methylcyclohexene (DL-limonene) by Inverse Gas Chromatography (IGC).
Assessment of the Thermodynamic Properties of DL-p-Mentha-1,8-diene, 4-Isopropyl-1-Methylcyclohexene (DL-limonene) by Inverse Gas Chromatography (IGC).
Journal of chromatographic science (2018-05-12)
Negin Farshchi, Ali Abbasian, Kambiz Larijani
RÉSUMÉ
Limonene is a colorless liquid hydrocarbon and had been investigated as a plasticizer for many plastics. Prediction of solubility between different materials is an advantage in many ways, one of the most convenient ways to know the compatibility of materials is to determine the degree of solubility of them in each other. The concept of "solubility parameter" can help practitioners in this way.In this study, inverse gas chromatography (IGC) method at infinite dilution was used for determination of the thermodynamic properties of DL-p-mentha-1,8-diene, 4-Isopropyl-1-methylcyclohexene (DL-limonene). The interaction between DL-limonene and 13 solvents were examined in the temperature range of 63-123°C through the assessment of the thermodynamic sorption parameters, the parameters of mixing at infinite dilution, the weight fraction activity coefficient and the Flory-Huggins interaction parameters. Additionally, the solubility parameter for DL-limonene and the temperature dependence of these parameters was investigated as well.Results show that there is a temperature dependence in solubility parameter, which increases by decreasing temperature. However, there were no specific dependence between interaction parameters and temperature, but chemical structure appeared to have a significant effect on them as well as on the type and strength of intermolecular interactions between DL-limonene and investigated solvents. The solubility parameter δ2 of DL-limonene determined to be 19.20 (J/cm3)0.5 at 25°C.