Merck
  • Down-regulation of transcription factor OVOL2 contributes to epithelial-mesenchymal transition in a noninvasive type of trophoblast implantation to the maternal endometrium.
Down-regulation of transcription factor OVOL2 contributes to epithelial-mesenchymal transition in a noninvasive type of trophoblast implantation to the maternal endometrium.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2018-02-03)
Rulan Bai, Kazuya Kusama, Keigo Nakamura, Toshihiro Sakurai, Koji Kimura, Atsushi Ideta, Yoshito Aoyagi, Kazuhiko Imakawa
RÉSUMÉ
Embryo implantation into the uterine endometrium is required for pregnancy establishment in most mammals. By using global expression analysis, we investigated the molecules that are related to epithelial-mesenchymal transition (EMT) in noninvasive bovine trophoblasts and found that the transcription factor, ovo-like zinc finger 2 ( OVOL2), which is essential for mesenchymal-epithelial transition in various cancers, was down-regulated after trophoblast attachment to the endometrial epithelium in utero. In cultured bovine trophoblast cells, OVOL2 down-regulation occurred only when cells were allowed to attach to bovine endometrial epithelial cells via the TEAD3/YAP signaling pathway. This resulted in the up-regulation of the EMT-associated transcription factors, ZEB1 and SNAI2, and the mesenchymal cell markers, N-cadherin ( CDH2) and vimentin ( VIM), whereas epithelial cell marker, E-cadherin ( CDH1), was down-regulated. In contrast, OVOL2 overexpression in bovine trophoblast cells exhibited a decrease in ZEB1 transcripts and an increase in E-cadherin. These observations revealed that ovo-like protein (OVOL)2 down-regulation occurred concurrently with conceptus implantation into the uterine endometrium via the YAP/TEAD3 signaling pathway, and suggest that the down-regulation of OVOL2 expression contributes to the up-regulation of EMT-related transcription factor expression, which enables EMT progression in the noninvasive bovine trophectoderm postimplantation.-Bai, R., Kusama, K., Nakamura, K., Sakurai, T., Kimura, K., Ideta, A., Aoyagi, Y., Imakawa, K. Down-regulation of transcription factor OVOL2 contributes to epithelial-mesenchymal transition in a noninvasive type of trophoblast implantation to the maternal endometrium.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
MISSION® esiRNA, targeting human NOTCH2