Key Documents
SYNF318100
Synthware recovery flask
100 mL, joint: ST/NS 29/42
Synonyme(s) :
Synthware F318100, oval shape flask
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.51
Matériaux
borosilicate glass flask
pear-shaped
single-neck
Joint
joint: ST/NS29/42
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F318100
Capacité
100 mL
100 mL
Description générale
Heavy wall design. All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Informations légales
Synthware is a trademark of Kemtech America Inc.