Critical Micelle Concentrations (CMCs) - Avanti® Polar Lipids
Definition
The critical micelle concentration (CMC) can be approximately defined as the lipid monomer concentration at which appreciable amounts (>5% of total) of micellar aggregates first begin to appear in the equilibrium: nM1<=>Mn
CMC Values
Products not available in the USA
Lipid Acronyms Key
PC = phosphatidylcholine
PG = phosphatidylglycerol
PS = phosphatidylserine
![Critical Micelle Concentrations (CMCs) - Avanti<sup>®</sup> Polar Lipids Lipid Acronyms Key](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/materials-science-and-engineering/drug-delivery/cmc-values-for-diacyl-phosphatidylcholine/cmc-values-for-diacyl-phosphatidylcholine.jpg)
CMC’s were determined in house by using a Pyrene Fluorescence technique or were obtained from the literature. Many of the literature values were confirmed by our in-house method.
The method is generally outlined by: Fluorescent Probes For Critical Micelle Concentration
References
1.
Derek M. 2013. Handbook of Lipid Bilayers. ISBN 0849332559. CRC Press, Inc..