HPLC Analysis of Yeast Lysate on Proteomix® SAX-NP3 Demonstrating High Separation Efficency
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column temp.
25 °C
column
Proteomix SAX-NP3, 5 cm x 4.6 mm I.D., 3 μm particles (Z777219)
detector
UV, 280 nm
flow rate
0.5 mL/min
gradient
0 - 100% B in 30 min
injection
40 μL
mobile phase
(A) 20 mM Tris, pH 9.0; (B) A + 0.5 M NaCl
sample
2.5 mg/mL
Description
Legal Information
Proteomix is a registered trademark of Sepax Technologies