HPLC Analysis of Hemoglobin HbA1C on Mono S® HR5/5
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Mono S HR5/5, 5 cm x 5 mm I.D., 10 μm (54808)
mobile phase
[A] 0.01M malonate-sodium hydroxide, pH 5.7; [B] 0.3M lithium chloride in mobile phase A
flow rate
2.0 mL/min
detector
VIS, 405 nm
injection
50 μL hemolysate, dilued 1:5
Description
Legal Information
Mono S is a registered trademark of Cytiva