HPLC Analysis of Fluroquinolone Antibiotics on Discovery® C18
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery C18, 15 cm × 4.6 mm I.D., 5 μm particles (504955)
mobile phase
[A] water (10 mM ammonium acetate, pH 3.0 with acetic acid); [B] acetonitrile; (85:15, A:B)
flow rate
1.0 mL/min
column temp.
35 °C
detector
UV, 254 nm
injection
10 μL
sample
50 μg/mL each in mobile phase
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany