HPLC Analysis of Flurbiprofen Enantiomers on Astec® Cellulose DMP
![HPLC Analysis of Flurbiprofen Enantiomers on Astec® Cellulose DMP application for HPLC](/deepweb/assets/sigmaaldrich/product/images/368/023/d73f0220-5177-401c-8a79-db252a162e33/800/d73f0220-5177-401c-8a79-db252a162e33.jpg)
Materials
analytical column
Product No.
Description
Pricing
mobile phase component
Product No.
Description
Pricing
standard
Product No.
Description
Pricing
CONDITIONS
column
Astec Cellulose DMP, 15 cm x 4.6 mm I.D., 5 μm particles (51098AST)
mobile phase
[A] heptane; [B] isopropanol; [C] trifluoroacetic acid (95:5:0.1, A:B:C)
flow rate
0.5 mL/min
column temp.
25 °C
detector
UV, 230 nm
injection
3 μL
sample
1 g/L in methanol