Merck
HPLC Analysis of Fat Soluble Vitamins on Ascentis® RP-Amide
HPLC Analysis of Fat Soluble Vitamins on Ascentis® RP-Amide application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® RP-Amide HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
standard
Product No.
Description
Pricing
(±)-α-Tocopherol
analytical standard
Vitamin D3 solution
1 mg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
CONDITIONS
column
Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 μm particles (565324-U)
mobile phase
acetonitrile
flow rate
2.0 mL/min
column temp.
35 °C
detector
UV, 215 nm
injection
20 μL
Description
Analysis Note
The Ascentis RP-Amide is shown to be suitable for the analysis of fat soluble vitamins, alpha tocopherol (vitamin E) and cholecalciferol (vitamin D3 ).
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany