HPLC Analysis of Deprenyl (Selegiline) Enantiomers on Astec® CYCLOBOND I 2000 DMP
![HPLC Analysis of Deprenyl (Selegiline) Enantiomers on Astec® CYCLOBOND I 2000 DMP application for HPLC](/deepweb/assets/sigmaaldrich/product/images/340/804/eb2daed8-bf21-413d-bee9-c44551e9fcfb/800/eb2daed8-bf21-413d-bee9-c44551e9fcfb.jpg)
CONDITIONS
column
CYCLOBOND I 2000 DMP, 25 cm x 4.6 mm I.D., 5 μm particles (20724AST)
mobile phase
[A] 20 mM ammonium acetate, pH 4.0; [B] methanol; (65:35, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
10 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null