HPLC Analysis of Coumafuryl Enantiomers on Astec® CHIROBIOTIC® V
![HPLC Analysis of Coumafuryl Enantiomers on Astec® CHIROBIOTIC® V application for HPLC](/deepweb/assets/sigmaaldrich/product/images/283/691/514dcfad-b8d2-44b4-bd7f-08367e775a3a/800/514dcfad-b8d2-44b4-bd7f-08367e775a3a.jpg)
CONDITIONS
column
CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 μm particles (11024AST)
mobile phase
[A] 20 mM ammonium nitrate, pH 5.5; [B] THF; (90:10, A:B)
flow rate
1.2 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
1 μL
sample
2 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC