Merck
GC Analysis of cis/trans Limonene Oxide Enantiomers on Astec® CHIRALDEX B-PH
GC Analysis of cis/trans Limonene Oxide Enantiomers on Astec® CHIRALDEX™ B-PH suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIRALDEX B-PH Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.12 μm
standard
Product No.
Description
Pricing
(R)-(+)-Limonene
analytical standard
(R)-(+)-Limonene
technical, ~90% (sum of enantiomers, GC)
(S)-(−)-Limonene
analytical standard
(S)-(−)-Limonene
96%
(R)-(+)-Limonene
97%
(S)-(−)-Limonene
≥95%, FG
(R)-(+)-Limonene
≥93%
CONDITIONS
column
Astec CHIRALDEX B-PH, 30 m x 0.25 mm I.D., 0.12 μm (71023AST)
oven
90 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 24 psi
injection
1 μL, 80:1 split
sample
3 mg/mL in methylene chloride
peak 1: d-cis or d-trans limonene oxide
peak 2: l-cis and trans limonene oxide
peak 3: d-trans or d-cis limonene oxide
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC